Assay Details
Target Gene Details
Entrez Gene ID: | 9837 |
Gene Name: | GINS complex subunit 1 |
Gene Aliases: |
PSF1 |
Location: |
Chr.20:25407470-25448568 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GINS1 | NM_021067.4 | NP_066545.3 | ||
| NR_134574.1 | ||||
| XM_017028162.1 | XP_016883651.1 | |||
| AK294593.1 | ||||
| BC012542.1 | AAH12542.1 | |||
| D80008.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1067379 | Chr.20:25374848 - 25612604 on Build GRCh38 | Gain |
|
| nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain |
|
| nsv585746 | Chr.20:25090818 - 25489420 on Build GRCh38 | Gain |
|
| dgv4290n100 | Chr.20:25086650 - 25494993 on Build GRCh38 | Gain |
|
| nsv1059494 | Chr.20:25396966 - 25461199 on Build GRCh38 | Gain |
|
| nsv833948 | Chr.20:25251090 - 25429904 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map