Assay Details
Target Gene Details
Entrez Gene ID: | 9777 |
Gene Name: | transmembrane 9 superfamily member 4 |
Gene Aliases: |
dJ836N17.2 |
Location: |
Chr.20:32109506-32167258 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM9SF4 | NM_014742.3 | NP_055557.2 | ||
| DC381660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833953 | Chr.20:32022592 - 32204778 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map