Assay Details
Target Gene Details
Entrez Gene ID: | 6780 |
Gene Name: | staufen double-stranded RNA binding protein 1 |
Gene Aliases: |
PPP1R150, STAU |
Location: |
Chr.20:49113339-49219295 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1067146 | Chr.20:48878894 - 49475527 on Build GRCh38 | Loss |
|
| esv2762079 | Chr.20:49095851 - 49155612 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78864916] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map