Assay Details
Target Gene Details
Entrez Gene ID: | 3172 |
Gene Name: | hepatocyte nuclear factor 4 alpha |
Gene Aliases: |
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14 |
Location: |
Chr.20:44355801-44432845 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNF4A | NM_001030003.2 | NP_001025174.1 | ||
| NM_001030004.2 | NP_001025175.1 | |||
| NM_001287182.1 | NP_001274111.1 | |||
| NM_001287183.1 | NP_001274112.1 | |||
| NM_001287184.1 | NP_001274113.1 | |||
| NM_175914.4 | NP_787110.2 | |||
| AK096973.1 | 1 | 1566 | ||
| AY680696.1 | AAT91237.1 | |||
| AY680697.1 | AAT91238.1 | |||
| AY680698.1 | AAT91239.1 | |||
| FJ608822.1 | ||||
| HQ692868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| nsv586065 | Chr.20:44261867 - 44505244 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map