Assay Details
Target Gene Details
Entrez Gene ID: | 101926888 |
Gene Name: | RALY antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:33992488-33994367 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RALY-AS1 | NR_109885.1 | |||
| NR_109886.1 | ||||
| AA447916.1 | ||||
| AA769840.1 |

Genomic Map