Assay Details
Target Gene Details
Entrez Gene ID: | 85508 |
Gene Name: | scratch family transcriptional repressor 2 |
Gene Aliases: |
ZNF898B |
Location: |
Chr.20:661596-676179 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCRT2 | NM_033129.3 | NP_149120.1 | ||
| AY129025.1 | AAM98768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644952 | Chr.20:386013 - 1119183 on Build GRCh38 | Gain |
|
| nsv1160671 | Chr.20:665086 - 674628 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115038969,rs74566866] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map