Assay Details
Target Gene Details
Entrez Gene ID: | 22913 |
Gene Name: | RALY heterogeneous nuclear ribonucleoprotein |
Gene Aliases: |
HNRPCL2, P542 |
Location: |
Chr.20:33993652-34084884 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RALY | NM_007367.3 | NP_031393.2 | ||
| NM_016732.2 | NP_057951.1 | |||
| XM_005260334.4 | XP_005260391.1 | |||
| XM_005260336.4 | XP_005260393.1 | |||
| XM_011528695.2 | XP_011526997.1 | |||
| XM_017027731.1 | XP_016883220.1 | |||
| AF148457.1 | AAF04487.1 | |||
| AK222915.1 | BAD96635.1 | |||
| AK226085.1 | ||||
| AK291054.1 | ||||
| AK291079.1 | ||||
| AK299766.1 | ||||
| BC103753.1 | AAI03754.1 | |||
| BC105018.1 | AAI05019.1 | |||
| BG751407.1 | ||||
| L38696.1 | AAC28898.1 |
More Information
Set Membership: |
|

Genomic Map