Assay Details
Target Gene Details
Entrez Gene ID: | 57158 |
Gene Name: | junctophilin 2 |
Gene Aliases: |
CMH17, JP-2, JP2 |
Location: |
Chr.20:44111697-44187578 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JPH2 | NM_020433.4 | NP_065166.2 | ||
| AK304863.1 | ||||
| BC043206.2 | AAH43206.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| nsv954043 | Chr.20:44109461 - 44117660 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs149534356] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map