Assay Details
Target Gene Details
Entrez Gene ID: | 140688 |
Gene Name: | nucleolar protein 4 like |
Gene Aliases: |
C20orf112, C20orf113, dJ1184F4.2, dJ1184F4.4 |
Location: |
Chr.20:32443059-32585073 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL4L | NM_001256798.1 | NP_001243727.1 | ||
| NM_080616.4 | NP_542183.2 | |||
| XM_005260282.4 | XP_005260339.1 | |||
| XM_005260283.4 | XP_005260340.1 | |||
| XM_005260284.4 | XP_005260341.1 | |||
| XM_005260285.4 | XP_005260342.2 | |||
| XM_005260286.4 | XP_005260343.1 | |||
| XM_005260288.2 | XP_005260345.1 | |||
| XM_005260289.4 | XP_005260346.1 | |||
| XM_006723702.3 | XP_006723765.1 | |||
| XM_006723703.3 | XP_006723766.1 | |||
| XM_011528562.2 | XP_011526864.1 | |||
| XM_011528563.1 | XP_011526865.1 | |||
| XM_017027669.1 | XP_016883158.1 | |||
| XM_017027670.1 | XP_016883159.1 | |||
| AK056286.1 | BAB71138.1 | |||
| BC065370.1 | AAH65370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509765 | Chr.20:32420379 - 32489248 on Build GRCh38 | Insertion |
|
| nsv954022 | Chr.20:32445698 - 32482397 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map