Assay Details
Target Gene Details
Entrez Gene ID: | 5126 |
Gene Name: | proprotein convertase subtilisin/kexin type 2 |
Gene Aliases: |
NEC 2, NEC-2, NEC2, PC2, SPC2 |
Location: |
Chr.20:17226107-17484578 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCSK2 | NM_001201528.1 | NP_001188457.1 | ||
| BC040546.1 | AAH40546.1 |
More Information
Set Membership: |
|

Genomic Map