Assay Details
Target Gene Details
Entrez Gene ID: | 140710 |
Gene Name: | suppressor of glucose, autophagy associated 1 |
Gene Aliases: |
C20orf117, KIAA0889, SOGA |
Location: |
Chr.20:36777442-36863684 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOGA1 | NM_080627.3 | 15 | 12467 | NP_542194.2 |
| NM_199181.2 | NP_954650.2 | |||
| AK126630.1 | BAC86621.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map