Assay Details
Target Gene Details
Entrez Gene ID: | 8839 |
Gene Name: | WNT1 inducible signaling pathway protein 2 |
Gene Aliases: |
CCN5, CT58, CTGF-L |
Location: |
Chr.20:44714204-44728041 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WISP2 | XM_017028117.1 | XP_016883606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458985 | Chr.20:44706933 - 44779789 on Build GRCh38 | Gain |
|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map