Assay Details
Target Gene Details
Entrez Gene ID: | 57506 |
Gene Name: | mitochondrial antiviral signaling protein |
Gene Aliases: |
CARDIF, IPS-1, IPS1, VISA |
Location: |
Chr.20:3846799-3876123 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAVS | NM_001206491.1 | NP_001193420.1 | ||
| NM_020746.4 | NP_065797.2 | |||
| NR_037921.1 | ||||
| AB033097.1 | ||||
| AB097003.1 | BAC77356.1 | |||
| AB232371.1 | BAE79738.1 | |||
| AK023799.1 | BAB14684.1 | |||
| AK123956.1 | BAC85734.1 | |||
| AK291785.1 | ||||
| AK296897.1 | ||||
| BC044952.1 | AAH44952.1 | |||
| BX649084.1 | ||||
| DQ167126.1 | ABA54890.1 | |||
| DQ174270.1 | AAZ80417.1 | |||
| DQ181928.1 | ABA19229.1 | |||
| EF467323.1 | ||||
| EF467324.1 | ||||
| KC415005.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645053 | Chr.20:3849918 - 3987827 on Build GRCh38 | Loss |
|
| nsv833899 | Chr.20:3709295 - 3878350 on Build GRCh38 | Loss |
|
| nsv527710 | Chr.20:3862380 - 3870321 on Build GRCh38 | Loss |
|
| esv2763216 | Chr.20:3858160 - 3879187 on Build GRCh38 | Loss |
|
| nsv828673 | Chr.20:3738382 - 4184811 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map