Assay Details
Target Gene Details
Entrez Gene ID: | 434 |
Gene Name: | agouti signaling protein |
Gene Aliases: |
AGSW, AGTI, AGTIL, ASP, SHEP9 |
Location: |
Chr.20:34186493-34269344 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASIP | XM_011528820.2 | XP_011527122.1 | ||
| XM_011528821.1 | XP_011527123.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3357 | Chr.20:34192245 - 34244945 on Build GRCh38 | Deletion |
|
| nsv1059756 | Chr.20:34143188 - 34368334 on Build GRCh38 | Gain |
|
| nsv510785 | Chr.20:34202450 - 34247215 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map