Assay Details
Target Gene Details
Entrez Gene ID: | 51605 |
Gene Name: | tRNA methyltransferase 6 |
Gene Aliases: |
CGI-09, GCD10, Gcd10p |
Location: |
Chr.20:5937832-5950558 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT6 | NM_001281467.1 | NP_001268396.1 | ||
| NM_015939.4 | NP_057023.2 | |||
| XM_017027875.1 | XP_016883364.1 | |||
| AB032979.1 | BAA86467.1 | |||
| AK000613.1 | ||||
| AK300812.1 | ||||
| BC001262.1 | AAH01262.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833905 | Chr.20:5774308 - 5941219 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73894087,rs76986775] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map