Assay Details
Target Gene Details
Entrez Gene ID: | 26164 |
Gene Name: | mitochondrial ribosome associated GTPase 2 |
Gene Aliases: |
GTPBP5, ObgH1, dJ1005F21.2 |
Location: |
Chr.20:62183025-62203568 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTG2 | NM_015666.3 | NP_056481.1 | ||
| XM_005260393.1 | XP_005260450.1 | |||
| XM_006723774.3 | XP_006723837.1 | |||
| XM_011528776.1 | XP_011527078.1 | |||
| XM_017027805.1 | XP_016883294.1 | |||
| XM_017027806.1 | XP_016883295.1 | |||
| XM_017027807.1 | XP_016883296.1 | |||
| XM_017027808.1 | XP_016883297.1 | |||
| AK001603.1 | BAA91783.1 | |||
| AK124572.1 | ||||
| AK299147.1 | ||||
| AK299189.1 | ||||
| AK299726.1 | ||||
| AK316027.1 | ||||
| AK316325.1 | ||||
| BC036716.2 | AAH36716.1 | |||
| CA976010.1 | ||||
| CB127579.1 | ||||
| DA970672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160689 | Chr.20:62188341 - 62304172 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map