Assay Details
Target Gene Details
Entrez Gene ID: | 343578 |
Gene Name: | Rho GTPase activating protein 40 |
Gene Aliases: |
C20orf95, dJ1100H13.4 |
Location: |
Chr.20:38601934-38650652 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP40 | NM_001164431.1 | NP_001157903.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058457 | Chr.20:38277492 - 38844429 on Build GRCh38 | Loss |
|
| nsv3372 | Chr.20:38587142 - 38619210 on Build GRCh38 | Insertion |
|
| esv1002417 | Chr.20:38609080 - 38611594 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map