Assay Details
Target Gene Details
Entrez Gene ID: | 117532 |
Gene Name: | transmembrane channel like 2 |
Gene Aliases: |
C20orf145 |
Location: |
Chr.20:2536607-2642185 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMC2 | NM_080751.2 | NP_542789.2 | ||
| AF417580.2 | AAL86401.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
| esv3644998 | Chr.20:2148722 - 2690694 on Build GRCh38 | Gain |
|
| dgv40e213 | Chr.20:2507648 - 2627511 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map