Assay Details
Target Gene Details
Entrez Gene ID: | 128637 |
Gene Name: | TBC1 domain family member 20 |
Gene Aliases: |
C20orf140, WARBM4 |
Location: |
Chr.20:435477-462553 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D20 | NM_144628.3 | NP_653229.1 | ||
| NR_111901.1 | ||||
| XM_005260661.1 | XP_005260718.1 | |||
| XM_006723540.3 | XP_006723603.1 | |||
| XM_017027645.1 | XP_016883134.1 | |||
| AB449906.1 | ||||
| AK095110.1 | ||||
| AK127062.1 | BAC86808.1 | |||
| AK291648.1 | ||||
| DA238706.1 | ||||
| DA311494.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066598 | Chr.20:422749 - 503206 on Build GRCh38 | Gain |
|
| esv3644953 | Chr.20:397290 - 502310 on Build GRCh38 | Gain |
|
| esv3644952 | Chr.20:386013 - 1119183 on Build GRCh38 | Gain |
|
| esv3644956 | Chr.20:405365 - 446917 on Build GRCh38 | Gain |
|
| nsv1056266 | Chr.20:401152 - 522755 on Build GRCh38 | Gain |
|
| nsv458842 | Chr.20:380145 - 444790 on Build GRCh38 | Loss |
|
| dgv4219n100 | Chr.20:80652 - 461103 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map