Assay Details
Target Gene Details
Entrez Gene ID: | 6638 |
Gene Name: | small nuclear ribonucleoprotein polypeptide N |
Gene Aliases: |
HCERN3, PWCR, RT-LI, SM-D, SMN, SNRNP-N, SNURF-SNRPN, sm-N |
Location: |
Chr.15:24823637-24978723 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRPN | NM_003097.4 | NP_003088.1 | ||
| NM_022805.3 | NP_073716.1 | |||
| NM_022806.3 | NP_073717.1 | |||
| NM_022807.3 | NP_073718.1 | |||
| NM_022808.3 | NP_073719.1 | |||
| AF319522.1 | ||||
| AF319523.1 | AAK18178.1 | |||
| AK123119.1 | 1 | 454 | ||
| AK222633.1 | BAD96353.1 | |||
| BC003180.1 | AAH03180.1 | |||
| BC024777.1 | AAH24777.1 | |||
| BC025178.1 | AAH25178.1 | |||
| DA131564.1 | ||||
| DA369087.1 | ||||
| J04615.1 | AAA36617.1 | |||
| L80004.1 | ||||
| L80005.1 | ||||
| U41303.1 | AAA98969.1 | |||
| X15892.1 | CAA33901.1 |
Target Gene Details
Entrez Gene ID: | 8926 |
Gene Name: | SNRPN upstream reading frame |
Gene Aliases: |
- |
Location: |
Chr.15:24954893-24978723 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNURF | NM_005678.4 | NP_005669.2 | ||
| AF101044.1 | AAD31391.1 | |||
| AK098563.1 | ||||
| DA134019.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045067 | Chr.15:24792359 - 25083568 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map