Assay Details
Target Gene Details
Entrez Gene ID: | 100132463 |
Gene Name: | claudin 24 |
Gene Aliases: |
CLDN21 |
Location: |
Chr.4:183321764-183322426 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLDN24 | NM_001185149.1 | 1 | 613 | NP_001172078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030766 | Chr.4:183305034 - 183948875 on Build GRCh38 | Gain |
|
| nsv1029647 | Chr.4:183268304 - 183372226 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map