Assay Details
Target Gene Details
Entrez Gene ID: | 285704 |
Gene Name: | repulsive guidance molecule family member b |
Gene Aliases: |
DRAGON |
Location: |
Chr.5:98768386-98796494 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGMB | NM_001012761.2 | NP_001012779.2 | ||
| XM_005271966.3 | XP_005272023.1 | |||
| XM_011543345.2 | XP_011541647.1 | |||
| XM_011543347.2 | XP_011541649.2 | |||
| XM_017009390.1 | XP_016864879.1 | |||
| XM_017009391.1 | XP_016864880.1 | |||
| AK074887.1 | BAC11268.1 | |||
| BC067736.1 | AAH67736.1 | |||
| BI763873.1 |
Target Gene Details
Entrez Gene ID: | 503569 |
Gene Name: | RGMB antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:98769618-98773084 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGMB-AS1 | NR_033932.1 | 1 | 340 | |
| AK093265.1 |
More Information
Set Membership: |
|

Genomic Map