Assay Details
Target Gene Details
Entrez Gene ID: | 54553 |
Gene Name: | uncharacterized protein DKFZP434I0714 |
Gene Aliases: |
- |
Location: |
Chr.4:152536264-152539263 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DKFZP434I0714 | NR_033797.2 | 1 | 857 | |
| AL137273.1 | 1 | 857 | CAB70671.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830116 | Chr.4:152415443 - 152613282 on Build GRCh38 | Gain |
|

Genomic Map