Assay Details
Target Gene Details
Entrez Gene ID: | 3835 |
Gene Name: | kinesin family member 22 |
Gene Aliases: |
A-328A3.2, KID, KNSL4, OBP, OBP-1, OBP-2, SEMDJL2 |
Location: |
Chr.16:29790062-29805385 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF22 | NM_001256269.1 | 7 | 916 | NP_001243198.1 |
| NM_001256270.1 | 6 | 1004 | NP_001243199.1 | |
| NM_007317.2 | 6 | 867 | NP_015556.1 | |
| AB017430.2 | 6 | 844 | BAA33019.2 | |
| AK223431.1 | 6 | 853 | BAD97151.1 | |
| AK294380.1 | 6 | 1243 | ||
| AK297893.1 | 5 | 540 | ||
| AK312234.1 | 6 | 858 | ||
| AK316050.1 | 6 | 1004 | ||
| AK316389.1 | 7 | 875 | ||
| BC004352.1 | 6 | 839 | AAH04352.1 | |
| BC028155.1 | 6 | 825 | AAH28155.1 | |
| BT007259.1 | 6 | 820 | AAP35923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833186 | Chr.16:29719552 - 29863674 on Build GRCh38 | Loss |
|
| dgv34n68 | Chr.16:29618081 - 29808034 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map