Assay Details
Target Gene Details
Entrez Gene ID: | 317762 |
Gene Name: | coiled-coil domain containing 85C |
Gene Aliases: |
- |
Location: |
Chr.14:99511266-99604390 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC85C | NM_001144995.1 | NP_001138467.1 | ||
| XM_011536705.2 | XP_011535007.1 | |||
| AK123840.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952236 | Chr.14:99504664 - 99604963 on Build GRCh38 | Deletion |
|
| esv3635481 | Chr.14:99277609 - 99600560 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map