Assay Details
Target Gene Details
Entrez Gene ID: | 4154 |
Gene Name: | muscleblind like splicing regulator 1 |
Gene Aliases: |
EXP, MBNL |
Location: |
Chr.3:152243656-152465780 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Gene Details
Entrez Gene ID: | 645843 |
Gene Name: | transmembrane protein 14E, pseudogene |
Gene Aliases: |
TMEM14E |
Location: |
Chr.3:152339698-152340990 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM14EP | NR_132656.1 | 1 | 144 | |
| AY358260.1 | 1 | 144 | AAQ88627.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483030 | Chr.3:152301789 - 152497220 on Build GRCh38 | Gain |
|
| nsv954493 | Chr.3:152196212 - 152409011 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map