Assay Details
Target Gene Details
Entrez Gene ID: | 9941 |
Gene Name: | endo/exonuclease (5'-3'), endonuclease G-like |
Gene Aliases: |
ENDOGL1, ENDOGL2, ENGL, ENGL-a, ENGL-b, ENGLA, ENGLB |
Location: |
Chr.3:38496127-38526305 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819153 | Chr.3:38513497 - 38517743 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map