Assay Details
Target Gene Details
Entrez Gene ID: | 23281 |
Gene Name: | microtubule associated tumor suppressor candidate 2 |
Gene Aliases: |
CAZIP, ICIS, KIAA0774, TIP150 |
Location: |
Chr.13:28820314-29505947 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTUS2 | NM_001033602.2 | NP_001028774.2 | ||
| NM_015233.5 | NP_056048.1 | |||
| XM_011535019.2 | XP_011533321.1 | |||
| XM_011535020.2 | XP_011533322.1 | |||
| XM_011535021.2 | XP_011533323.1 | |||
| XM_011535022.2 | XP_011533324.1 | |||
| XM_017020502.1 | XP_016875991.1 | |||
| AB018317.2 | ||||
| AK293970.1 | ||||
| AK302516.1 | ||||
| BC032481.1 | AAH32481.1 | |||
| BC119654.1 | ||||
| BC119655.1 | ||||
| BC129985.1 | ||||
| BC150245.1 | ||||
| BM906724.1 |
Target Gene Details
Entrez Gene ID: | 100874107 |
Gene Name: | MTUS2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:29476515-29487750 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTUS2-AS1 | NR_046378.1 | |||
| BI223246.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs139322757] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map