Assay Details
Target Gene Details
Entrez Gene ID: | 1794 |
Gene Name: | dedicator of cytokinesis 2 |
Gene Aliases: |
IMD40 |
Location: |
Chr.5:169637247-170083382 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK2 | NM_004946.2 | NP_004937.1 | ||
| XM_011534449.2 | XP_011532751.1 | |||
| XM_011534450.2 | XP_011532752.1 | |||
| XM_011534451.2 | XP_011532753.1 | |||
| XM_017009189.1 | XP_016864678.1 | |||
| XM_017009190.1 | XP_016864679.1 | |||
| AK128107.1 | ||||
| BC016996.1 | AAH16996.1 | |||
| BC104900.1 | AAI04901.1 | |||
| BC113457.1 | ||||
| D86964.1 |
Target Gene Details
Entrez Gene ID: | 100131897 |
Gene Name: | family with sequence similarity 196 member B |
Gene Aliases: |
C5orf57 |
Location: |
Chr.5:169861304-169980740 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM196B | NM_001129891.1 | NP_001123363.1 | ||
| XM_011534415.2 | XP_011532717.1 | |||
| XM_017008908.1 | XP_016864397.1 | |||
| AK055998.1 |
More Information
Set Membership: |
|

Genomic Map