Assay Details
Target Gene Details
Entrez Gene ID: | 101927100 |
Gene Name: | uncharacterized LOC101927100 |
Gene Aliases: |
- |
Location: |
Chr.5:115602155-115620419 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927100 | NR_109874.1 | |||
| NR_109875.1 | ||||
| AW592839.1 | ||||
| DA313138.1 |
Target Gene Details
Entrez Gene ID: | 100302736 |
Gene Name: | TMED7-TICAM2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.5:115578496-115626179 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED7-TICAM2 | NM_001164468.3 | NP_001157940.1 | ||
| NM_001164469.3 | NP_001157941.1 | |||
| AK075218.1 | BAC11479.1 | |||
| AY275836.1 | AAP81748.1 | |||
| AY304581.1 | AAQ97430.1 | |||
| AY304583.1 | AAQ97432.1 | |||
| AY304584.1 | AAQ97433.1 |
More Information
Set Membership: |
|

Genomic Map