Assay Details
Target Gene Details
Entrez Gene ID: | 26064 |
Gene Name: | retinoic acid induced 14 |
Gene Aliases: |
NORPEG, RAI13 |
Location: |
Chr.5:34656328-34832612 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAI14 | NM_001145520.1 | NP_001138992.1 | ||
| NM_001145522.1 | NP_001138994.1 | |||
| NM_015577.2 | NP_056392.2 | |||
| XM_011514024.1 | XP_011512326.1 | |||
| XM_017009335.1 | XP_016864824.1 | |||
| XM_017009336.1 | XP_016864825.1 | |||
| AB037755.1 | ||||
| AF155135.1 | AAF44722.1 | |||
| AK314379.1 | ||||
| BC052988.1 | AAH52988.1 | |||
| DA485361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1090e214 | Chr.5:34558763 - 34661785 on Build GRCh38 | Gain |
|
| dgv9720n54 | Chr.5:34656423 - 34657292 on Build GRCh38 | Loss |
|
| nsv4786 | Chr.5:34655988 - 34684200 on Build GRCh38 | Insertion |
|
| nsv519240 | Chr.5:33950598 - 34749311 on Build GRCh38 | Gain |
|
| dgv9717n54 | Chr.5:34656185 - 34657239 on Build GRCh38 | Gain+Loss |
|
| esv24799 | Chr.5:34656174 - 34657316 on Build GRCh38 | Loss |
|
| esv3604679 | Chr.5:34492621 - 34936206 on Build GRCh38 | Gain |
|
| nsv830260 | Chr.5:34544620 - 34764299 on Build GRCh38 | Gain |
|

Genomic Map