Assay Details
Target Gene Details
Entrez Gene ID: | 414201 |
Gene Name: | ZNF32 antisense RNA 3 |
Gene Aliases: |
C10orf44, ZNF32-OT1, bA402L1.3 |
Location: |
Chr.10:43628817-43674699 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF32-AS3 | NR_038867.1 | 2 | 441 |

Genomic Map