Assay Details
Target Gene Details
Entrez Gene ID: | 100133315 |
Gene Name: | transient receptor potential cation channel, subfamily C, member 2-like |
Gene Aliases: |
- |
Location: |
Chr.11:71865509-71928447 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100133315 | NR_029192.1 | |||
| BQ014444.1 | ||||
| BX102226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759836 | Chr.11:71652593 - 71946530 on Build GRCh38 | Gain+Loss |
|
| nsv383 | Chr.11:71898560 - 71916281 on Build GRCh38 | Insertion |
|
| nsv509428 | Chr.11:71875077 - 71937297 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map