Assay Details
Target Gene Details
Entrez Gene ID: | 117195 |
Gene Name: | MAS related GPR family member X3 |
Gene Aliases: |
GPCR, MRGX3, SNSR1, SNSR2 |
Location: |
Chr.11:18120955-18138480 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRX3 | NM_054031.3 | NP_473372.3 | ||
| XM_011519882.2 | XP_011518184.1 | |||
| BC067292.1 | AAH67292.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23532 | Chr.11:18117115 - 18179170 on Build GRCh38 | Gain |
|
| esv2744166 | Chr.11:18129836 - 18177209 on Build GRCh38 | Deletion |
|
| nsv971990 | Chr.11:18132374 - 18143365 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map