Assay Details
Target Gene Details
Entrez Gene ID: | 643338 |
Gene Name: | chromosome 15 open reading frame 62 |
Gene Aliases: |
- |
Location: |
Chr.15:40769961-40772449 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C15orf62 | NM_001130448.2 | 1 | 318 | NP_001123920.1 |
| AK291344.1 | 1 | 200 | ||
| DA761028.1 | 1 | 318 |
Target Gene Details
Entrez Gene ID: | 55192 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C17 |
Gene Aliases: |
- |
Location: |
Chr.15:40767448-40807478 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC17 | NM_018163.2 | NP_060633.1 | ||
| XM_017022401.1 | XP_016877890.1 | |||
| AK001496.1 | BAA91724.1 | |||
| BC000048.2 | AAH00048.1 |
More Information
Set Membership: |
|

Genomic Map