Assay Details
Target Gene Details
Entrez Gene ID: | 642273 |
Gene Name: | family with sequence similarity 110 member C |
Gene Aliases: |
- |
Location: |
Chr.2:38814-47077 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM110C | NM_001077710.2 | NP_001071178.2 | ||
| XM_011510372.2 | XP_011508674.1 | |||
| XM_017004691.1 | XP_016860180.1 | |||
| AL137761.1 | ||||
| DQ431183.1 | ABD92775.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011864 | Chr.2:12772 - 132364 on Build GRCh38 | Loss |
|
| esv2751887 | Chr.2:12994 - 251673 on Build GRCh38 | Loss |
|
| nsv1013177 | Chr.2:33889 - 206851 on Build GRCh38 | Gain |
|
| esv3589587 | Chr.2:23163 - 99614 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map