Assay Details
Target Gene Details
Entrez Gene ID: | 2034 |
Gene Name: | endothelial PAS domain protein 1 |
Gene Aliases: |
ECYT4, HIF2A, HLF, MOP2, PASD2, bHLHe73 |
Location: |
Chr.2:46297402-46386703 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPAS1 | NM_001430.4 | NP_001421.2 | ||
| BC051338.1 | AAH51338.1 | |||
| CD109908.1 | ||||
| U51626.1 | AAC51212.1 | |||
| U81984.1 | AAB41495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv581743 | Chr.2:46272597 - 46310465 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map