Assay Details
Target Gene Details
Entrez Gene ID: | 284992 |
Gene Name: | coiled-coil domain containing 150 |
Gene Aliases: |
- |
Location: |
Chr.2:196639632-196733668 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC150 | NM_001080539.1 | NP_001074008.1 | ||
| XM_006712438.3 | XP_006712501.1 | |||
| XM_011510985.2 | XP_011509287.1 | |||
| XM_011510987.2 | XP_011509289.1 | |||
| XM_011510988.2 | XP_011509290.1 | |||
| XM_017003861.1 | XP_016859350.1 | |||
| XM_017003862.1 | XP_016859351.1 | |||
| XM_017003863.1 | XP_016859352.1 | |||
| XM_017003864.1 | XP_016859353.1 | |||
| XM_017003865.1 | XP_016859354.1 | |||
| XM_017003866.1 | XP_016859355.1 | |||
| AK301665.1 | ||||
| AK302685.1 |
Target Gene Details
Entrez Gene ID: | 100130452 |
Gene Name: | uncharacterized LOC100130452 |
Gene Aliases: |
- |
Location: |
Chr.2:196700634-196713012 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130452 | NR_034036.1 | |||
| AI239495.1 | ||||
| BX103183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575323 | Chr.2:196674745 - 197095661 on Build GRCh38 | Gain |
|
| nsv997576 | Chr.2:196690505 - 196735570 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map