Assay Details
Target Gene Details
Entrez Gene ID: | 51011 |
Gene Name: | fumarylacetoacetate hydrolase domain containing 2A |
Gene Aliases: |
CGI-105 |
Location: |
Chr.2:95402691-95416702 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAHD2A | XM_011511282.2 | XP_011509584.1 | ||
| XM_011511283.2 | XP_011509585.1 | |||
| XM_011511284.2 | XP_011509586.1 | |||
| XM_011511285.2 | XP_011509587.1 | |||
| XM_011511286.2 | XP_011509588.1 | |||
| XM_011511287.2 | XP_011509589.1 | |||
| XM_011511288.2 | 9 | 1962 | XP_011509590.1 | |
| XM_011511289.2 | 9 | 3031 | XP_011509591.1 | |
| XM_017004236.1 | 10 | 2408 | XP_016859725.1 | |
| XM_017004237.1 | 10 | 2537 | XP_016859726.1 | |
| XM_017004238.1 | 10 | 2405 | XP_016859727.1 | |
| XM_017004239.1 | 9 | 2291 | XP_016859728.1 | |
| XM_017004240.1 | 9 | 2402 | XP_016859729.1 | |
| XM_017004241.1 | 7 | 2146 | XP_016859730.1 | |
| XM_017004242.1 | 7 | 1406 | XP_016859731.1 | |
| XM_017004245.1 | 6 | 1983 | XP_016859734.1 | |
| XM_017004246.1 | 7 | 1845 | XP_016859735.1 | |
| XM_017004247.1 | 6 | 1243 | XP_016859736.1 | |
| XM_017004248.1 | 6 | 1354 | XP_016859737.1 | |
| XM_017004249.1 | 5 | 1174 | XP_016859738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160825 | Chr.2:95050220 - 95445574 on Build GRCh38 | Duplication |
|
| esv2759076 | Chr.2:95304390 - 95486811 on Build GRCh38 | Loss |
|
| nsv998014 | Chr.2:95247530 - 95630882 on Build GRCh38 | Gain |
|
| nsv961449 | Chr.2:95403429 - 95415505 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map