Assay Details
Target Gene Details
Entrez Gene ID: | 199974 |
Gene Name: | cytochrome P450 family 4 subfamily Z member 1 |
Gene Aliases: |
CYP4A20 |
Location: |
Chr.1:47067488-47118320 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4Z1 | NM_178134.2 | NP_835235.1 | ||
| AK292175.1 | ||||
| AY262056.1 | AAO89257.1 | |||
| AY358631.1 | AAQ88994.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585951 | Chr.1:47080141 - 47121317 on Build GRCh38 | Gain |
|
| esv24824 | Chr.1:47099918 - 47108510 on Build GRCh38 | Loss |
|
| esv2763300 | Chr.1:46892860 - 47163102 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map