Assay Details
Target Gene Details
Entrez Gene ID: | 642587 |
Gene Name: | MIR205 host gene |
Gene Aliases: |
LINC00510 |
Location: |
Chr.1:209428820-209432552 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR205HG | NM_001104548.1 | 1 | 36 | NP_001098018.1 |
| XM_017002058.1 | 1 | 39 | XP_016857547.1 | |
| AK091113.1 | 1 | 39 | ||
| BG674039.1 | ||||
| CV575609.1 | ||||
| DA461662.1 | 1 | 36 |

Genomic Map