Assay Details
Target Gene Details
Entrez Gene ID: | 642587 |
Gene Name: | MIR205 host gene |
Gene Aliases: |
LINC00510 |
Location: |
Chr.1:209428820-209432552 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR205HG | NM_001104548.1 | NP_001098018.1 | ||
| XM_017002058.1 | XP_016857547.1 | |||
| AK091113.1 | 1 | 2060 | ||
| AW275825.1 | ||||
| BG674039.1 | ||||
| BM980083.1 | ||||
| CV575609.1 |

Genomic Map