Assay Details
Target Gene Details
Entrez Gene ID: | 3775 |
Gene Name: | potassium two pore domain channel subfamily K member 1 |
Gene Aliases: |
DPK, HOHO, K2P1, K2p1.1, KCNO1, TWIK-1, TWIK1 |
Location: |
Chr.1:233614004-233672512 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK1 | NM_002245.3 | NP_002236.1 | ||
| XM_011544184.2 | 1 | 52 | XP_011542486.1 | |
| AK090902.1 | 1 | 52 | ||
| AK314016.1 | ||||
| BC018051.1 | AAH18051.1 | |||
| U33632.1 | AAB01688.1 | |||
| U76996.1 | AAB97878.1 | |||
| U90065.1 | AAB51147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528071 | Chr.1:233620319 - 233629781 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map