Assay Details
Target Gene Details
Entrez Gene ID: | 390874 |
Gene Name: | one cut homeobox 3 |
Gene Aliases: |
OC3 |
Location: |
Chr.19:1753663-1775445 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ONECUT3 | NM_001080488.1 | NP_001073957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss |
|
| esv2671081 | Chr.19:1746946 - 1765393 on Build GRCh38 | Deletion |
|
| nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map