Assay Details
Target Gene Details
Entrez Gene ID: | 337985 |
Gene Name: | keratin associated protein 20-3 |
Gene Aliases: |
KAP19D, KAP20.3, KRTAP19P4 |
Location: |
Chr.21:30642864-30643136 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP20-3 | NM_001128077.1 | 1 | 194 | NP_001121549.1 |
| AB096958.1 | 1 | 194 | BAE46373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057467 | Chr.21:30318003 - 30993921 on Build GRCh38 | Gain |
|

Genomic Map