Assay Details
Target Gene Details
Entrez Gene ID: | 100131902 |
Gene Name: | keratin associated protein 25-1 |
Gene Aliases: |
KAP25.1 |
Location: |
Chr.21:30289145-30289514 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP25-1 | NM_001128598.1 | 1 | 290 | NP_001122070.1 |
| AB180043.1 | 1 | 290 | BAE46382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834076 | Chr.21:30216502 - 30383428 on Build GRCh38 | Gain |
|
| nsv587355 | Chr.21:30185998 - 30289834 on Build GRCh38 | Loss |
|
| nsv587354 | Chr.21:29978351 - 30620545 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map