Assay Details
Target Gene Details
Entrez Gene ID: | 100861412 |
Gene Name: | fibrinogen silencer binding protein |
Gene Aliases: |
- |
Location: |
Chr.8:94427712-94436952 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSBP | NM_001256141.1 | 2 | 906 | NP_001243070.1 |
| AF007866.1 | 2 | 858 | AAD09313.1 |
Target Gene Details
Entrez Gene ID: | 25788 |
Gene Name: | RAD54 homolog B (S. cerevisiae) |
Gene Aliases: |
RDH54 |
Location: |
Chr.8:94371960-94475115 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD54B | NM_001205262.2 | 4 | 883 | NP_001192191.1 |
| NM_001205263.1 | NP_001192192.1 | |||
| NM_012415.3 | NP_036547.1 | |||
| AF112481.1 | AAD34331.1 | |||
| AK290437.1 | ||||
| AK307516.1 | ||||
| BC001965.2 | AAH01965.1 | |||
| BC033710.2 | 3 | 768 | AAH33710.2 | |
| BF766767.1 | ||||
| BP872374.1 | ||||
| DA553267.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6313 | Chr.8:94412057 - 94445643 on Build GRCh38 | Insertion |
|
| nsv1031480 | Chr.8:94363468 - 94435179 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map