Assay Details
Target Gene Details
Entrez Gene ID: | 55074 |
Gene Name: | oxidation resistance 1 |
Gene Aliases: |
Nbla00307, TLDC3 |
Location: |
Chr.8:106270178-106752694 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXR1 | NM_001198533.1 | NP_001185462.1 | ||
| NM_018002.3 | NP_060472.2 | |||
| XM_017013596.1 | XP_016869085.1 | |||
| AK124441.1 | BAC85852.1 | |||
| DA232248.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv29304 | Chr.8:106270637 - 106271462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map