Assay Details
Target Gene Details
Entrez Gene ID: | 100132813 |
Gene Name: | uncharacterized LOC100132813 |
Gene Aliases: |
- |
Location: |
Chr.8:109644115-109648084 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100132813 | NM_001317989.1 | 2 | 146 | NP_001304918.1 |
| AK093952.1 | 2 | 147 |
Target Gene Details
Entrez Gene ID: | 55638 |
Gene Name: | syntabulin |
Gene Aliases: |
GOLSYN, OCSYN, SNPHL |
Location: |
Chr.8:109574176-109691791 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
More Information
Additional Information:
For this assay, SNP(s) [rs77335041] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map