Assay Details
Target Gene Details
Entrez Gene ID: | 100132813 |
Gene Name: | uncharacterized LOC100132813 |
Gene Aliases: |
- |
Location: |
Chr.8:109644115-109648084 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100132813 | NM_001317989.1 | 2 | 956 | NP_001304918.1 |
| AK093952.1 | 2 | 957 |
Target Gene Details
Entrez Gene ID: | 55638 |
Gene Name: | syntabulin |
Gene Aliases: |
GOLSYN, OCSYN, SNPHL |
Location: |
Chr.8:109574176-109691791 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYBU | NM_001099743.1 | NP_001093213.1 | ||
| NM_001099744.1 | NP_001093214.1 | |||
| NM_001099745.1 | NP_001093215.1 | |||
| NM_001099746.1 | NP_001093216.1 | |||
| NM_001099747.1 | NP_001093217.1 | |||
| NM_001099748.1 | NP_001093218.1 | |||
| NM_001099749.1 | NP_001093219.1 | |||
| NM_001099750.1 | NP_001093220.1 | |||
| NM_017786.5 | NP_060256.3 | |||
| XM_017013615.1 | XP_016869104.1 | |||
| AB195676.1 | BAD72976.1 | |||
| AB195677.1 | BAD72977.1 | |||
| AB195678.1 | BAD72978.1 | |||
| AK000373.1 | BAA91121.1 | |||
| AK290469.1 | ||||
| AL713645.1 | CAD28457.2 | |||
| BG702715.1 | ||||
| CA395128.1 | ||||
| DA155137.1 | ||||
| DA530258.1 | ||||
| DA803330.1 | ||||
| DB155885.1 | ||||
| DC350594.1 | ||||
| DN993094.1 |
More Information
Set Membership: |
|

Genomic Map